Methyl-2-(6-bromo-2H-1,3-benzodioxol-5-yl)acetate ≥95%

Supplier: Matrix Scientific

064089-500MG 064089-1G
101927-020EA 803.08 USD
101927-020 101927-022
Methyl-2-(6-bromo-2H-1,3-benzodioxol-5-yl)acetate ≥95%
Methyl-2-(6-bromo-2H-1,3-benzodioxol-5-yl)acetate

Hazard: Irritant

Formula: C₁₀H₉BrO₄
Melting Pt: 83…85 °C
MDL Number: MFCD09865161
CAS Number: 51665-84-0

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR