Ethyl-4-{[(pyridin-3-yl)carbamothioyl]amino}benzoate ≥95%

Supplier: Matrix Scientific

064085-500MG 064085-1G
101927-006EA 298.46 USD
101927-006 101927-008
Ethyl-4-{[(pyridin-3-yl)carbamothioyl]amino}benzoate ≥95%
Ethyl-4-{[(pyridin-3-yl)carbamothioyl]amino}benzoate

Hazard: Irritant

Formula: C₁₅H₁₅N₃O₂S
MW: 301.37 g/mol
Melting Pt: 135…136 °C
MDL Number: MFCD04297579
CAS Number: 725275-38-7

Order Now

Specification Test Results

Melting point 135-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR