Ethyl-4-{[(2,3-dihydro-1H-inden-5-yl)carbamoyl]amino}benzoate ≥95%
Supplier: Matrix Scientific
064082-500MG
064082-1G
101926-994EA
193.85
USD
101926-994
101926-996
Ethyl-4-{[(2,3-dihydro-1H-inden-5-yl)carbamoyl]amino}benzoate ≥95%
Ethyl-4-{[(2,3-dihydro-1H-inden-5-yl)carbamoyl]amino}benzoate
Hazard: Irritant
Formula:
C₁₉H₂₀N₂O₃ Melting Pt: 212…215 °C |
MDL Number:
MFCD06105406 CAS Number: 893009-73-9 |
Specification Test Results
Melting point | 212-215°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...