Ethyl-4-[(cyclohexylcarbamoyl)amino]benzoate ≥95%

Supplier: Matrix Scientific

064081-500MG 064081-1G
101926-990EA 193.85 USD
101926-990 101926-992
Ethyl-4-[(cyclohexylcarbamoyl)amino]benzoate ≥95%
Ethyl-4-[(cyclohexylcarbamoyl)amino]benzoate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
Melting Pt: 169…171 °C
MDL Number: MFCD01472406
CAS Number: 544438-17-7

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR