Ethyl-5-cyano-2-hydroxy-6-methylpyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

064080-500MG 064080-1G 064080-5G
101926-984EA 193.85 USD
101926-984 101926-986 101926-988
Ethyl-5-cyano-2-hydroxy-6-methylpyridine-3-carboxylate ≥95%
Ethyl-5-cyano-2-hydroxy-6-methylpyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃
Melting Pt: 216…218 °C
MDL Number: MFCD01311996
CAS Number: 75894-42-7

Order Now

Specification Test Results

Melting point 216-218°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR