5,6-Dimethoxy-2-nitroso-2,3-dihydro-1H-inden-1-one ≥95%

Supplier: Matrix Scientific

064079-500MG 064079-1G
101926-980EA 298.46 USD
101926-980 101926-982
5,6-Dimethoxy-2-nitroso-2,3-dihydro-1H-inden-1-one ≥95%
5,6-Dimethoxy-2-nitroso-2,3-dihydro-1H-inden-1-one

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
Melting Pt: 217…219 °C
MDL Number: MFCD18432057
CAS Number: 2107-85-9

Order Now

Specification Test Results

Melting point 217-219°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR