6-Chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

064077-500MG 064077-1G
101926-972EA 675.38 USD
101926-972 101926-974
6-Chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylic acid ≥95%
6-Chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₅ClN₂O₂
Melting Pt: 350 °C+
MDL Number: MFCD09878691
CAS Number: 800401-54-1

Order Now

Specification Test Results

Melting point 350°C+

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR