5-Bromo-2-fluoronicotinic acid ≥95%
Supplier: Matrix Scientific
064072-1G
064072-5G
101926-954EA
307.69
USD
101926-954
102551-644
5-Bromo-2-fluoronicotinic acid ≥95%
5-Bromo-2-fluoronicotinic acid
Hazard: Irritant
Formula:
C₆H₃BrFNO₂ Melting Pt: 142…144 °C |
MDL Number:
MFCD06797939 CAS Number: 29241-66-5 |
Specification Test Results
Melting point | 142-144°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...