4-Bromo-2-(bromomethyl)-1-benzothiophene ≥95%

Supplier: Matrix Scientific

064069-500MG 064069-1G
101926-940EA 803.08 USD
101926-940 101926-942
4-Bromo-2-(bromomethyl)-1-benzothiophene ≥95%
4-Bromo-2-(bromomethyl)-1-benzothiophene

Hazard: Irritant

Formula: C₉H₆Br₂S
Melting Pt: 95…97 °C
MDL Number: MFCD19381764
CAS Number: 1312118-04-9

Order Now

Specification Test Results

Melting point 95-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR