4-Bromo-1-benzothiophene-2-carbonitrile ≥95%

Supplier: Matrix Scientific

064067-500MG 064067-1G
101926-932EA 803.08 USD
101926-932 101926-934
4-Bromo-1-benzothiophene-2-carbonitrile ≥95%
4-Bromo-1-benzothiophene-2-carbonitrile

Hazard: Irritant

Formula: C₉H₄BrNS
Melting Pt: 121…122 °C
MDL Number: MFCD19381765
CAS Number: 1312118-13-0

Order Now

Specification Test Results

Melting point 121-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR