4-Bromo-1-benzothiophene-2-carboxamide ≥95%

Supplier: Matrix Scientific

064065-500MG 064065-1G
101926-924EA 467.69 USD
101926-924 101926-926
4-Bromo-1-benzothiophene-2-carboxamide ≥95%
4-Bromo-1-benzothiophene-2-carboxamide

Hazard: Irritant

Formula: C₉H₆BrNOS
Melting Pt: 182…184 °C
MDL Number: MFCD19381740
CAS Number: 93103-86-7

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR