1-Ethyl-3-phenyl-2-thiourea

Supplier: Matrix Scientific

057077-10G 057077-50G
101912-280EA 126.15 USD
101912-280 101912-282
1-Ethyl-3-phenyl-2-thiourea
1-Ethyl-3-phenyl-2-thiourea

TCA listed: T
Hazard: Irritant

Formula: C₉H₁₂N₂S
Melting Pt: 68…69 °C
MDL Number: MFCD00022133
CAS Number: 2741-06-2

Order Now

Specification Test Results

Melting point 102-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR