3-(3-Ethoxy-4-methoxyphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

057065-5G 057065-25G
101912-230EA 206.15 USD
101912-230 101912-232
3-(3-Ethoxy-4-methoxyphenyl)propionic acid ≥98%
3-(3-Ethoxy-4-methoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₄
Melting Pt: 103…105 °C
MDL Number: MFCD09025689
CAS Number: 31282-89-0

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR