1,3-Di(p-tolyl)-2-thiourea ≥99%

Supplier: Matrix Scientific

057055-10G 057055-50G
101912-198EA 167.69 USD
101912-198 101912-200
1,3-Di(p-tolyl)-2-thiourea ≥99%
1,3-Di(p-tolyl)-2-thiourea

Hazard: Irritant

Formula: C₁₅H₁₆N₂S
Melting Pt: 182…184 °C
MDL Number: MFCD00008541
CAS Number: 621-01-2

Order Now

Specification Test Results

Melting point 182-184°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR