1,3-Di-4-piperidylpropane

Supplier: Matrix Scientific

Synonyms: 1,3-Bis(4-piperidyl)propane, 4,4'-Trimethylenedipiperidine, 1,3-Di(piperidin-4-yl)propane

057053-10G 057053-50G
101912-190EA 167.69 USD
101912-190 101912-192
1,3-Di-4-piperidylpropane
1,3-Di-4-piperidylpropane

Hazard: Irritant

Formula: C₁₃H₂₆N₂
Melting Pt: 66 °C
MDL Number: MFCD00038013
CAS Number: 16898-52-5

Order Now

Specification Test Results

Melting point 66°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR