4-Hydroxy-2,6-dimethylbenzaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dimethyl-4-hydroxybenzaldehyde

057036-5G 057036-25G 057036-100G
101912-126EA 36.92 USD
101912-126 101912-128 102833-198
4-Hydroxy-2,6-dimethylbenzaldehyde ≥98%
4-Hydroxy-2,6-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.1700 g/mol
Melting Pt: 193-195°C
MDL Number: MFCD00016979
CAS Number: 70547-87-4

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR