4,4'-Diisothiocyanatodiphenylmethane ≥98%

Supplier: Matrix Scientific

057023-5G 057023-25G
101912-080EA 161.54 USD
101912-080 101912-082
4,4'-Diisothiocyanatodiphenylmethane ≥98%
4,4'-Diisothiocyanatodiphenylmethane

Hazard: Irritant

Formula: C₁₅H₁₀N₂S₂
Melting Pt: 137…139 °C
MDL Number: MFCD01310811
CAS Number: 2798-05-2

Order Now

Specification Test Results

Melting point 137-139°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR