3-(2,3-Dichlorophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

057010-5G 057010-25G
101912-034EA 115.38 USD
101912-034 101912-036
3-(2,3-Dichlorophenyl)propionic acid ≥98%
3-(2,3-Dichlorophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₈Cl₂O₂
Melting Pt: 116…118 °C
MDL Number: MFCD06823953
CAS Number: 57915-79-4

Order Now

Specification Test Results

Melting point 116-118°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR