1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea

Supplier: Matrix Scientific

056986-2G 056986-10G
101911-968EA 243.08 USD
101911-968 101911-970
1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea
1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea

Hazard: Irritant

Formula: C₈H₆ClF₃N₂S
MW: 254.66 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD00041182
CAS Number: 207919-03-7

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR