4-(4-Chlorophenyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

Synonyms: N-(4-Chlorophenyl)hydrazinecarbothioamide

056982-5G 056982-25G
101911-952EA 140 USD
101911-952 101911-954
4-(4-Chlorophenyl)-3-thiosemicarbazide
4-(4-Chlorophenyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₇H₈ClN₃S
MW: 201.68 g/mol
Melting Pt: 177-178°C (dec)
MDL Number: MFCD00041295
CAS Number: 22814-92-2

Order Now

Specification Test Results

Melting point 177-178°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR