4-(2-Chlorophenyl)-3-thiosemicarbazide
Supplier: Matrix Scientific
056981-5G
056981-25G
101911-948EA
156.92
USD
101911-948
101911-950
4-(2-Chlorophenyl)-3-thiosemicarbazide
4-(2-Chlorophenyl)-3-thiosemicarbazide
Hazard: Irritant
Formula:
C₇H₈ClN₃S Melting Pt: 136-138°C |
MDL Number:
MFCD00041271 CAS Number: 42135-75-1 |
Specification Test Results
Melting point | 136-138°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...