2-Chloro-4-nitrobenzonitrile ≥98%

Supplier: Matrix Scientific

056974-5G 056974-25G
101911-920EA 183.08 USD
101911-920 101911-922
2-Chloro-4-nitrobenzonitrile ≥98%
2-Chloro-4-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃ClN₂O₂
Melting Pt: 78…80 °C
MDL Number: MFCD00052203
CAS Number: 28163-00-0

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR