4-Chloro-3-methylbenzhydrazide

Supplier: Matrix Scientific

056965-1G 056965-10G
101911-896EA 98.46 USD
101911-896 101911-898
4-Chloro-3-methylbenzhydrazide
4-Chloro-3-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₉ClN₂O
Melting Pt: 164…166 °C
MDL Number: MFCD03425683
CAS Number: 864178-66-5

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR