4-(4-Chlorobenzyl)-3-thiosemicarbazide
Supplier: Matrix Scientific
056950-2G
056950-10G
101911-842EA
172.31
USD
101911-842
101911-844
4-(4-Chlorobenzyl)-3-thiosemicarbazide
4-(4-Chlorobenzyl)-3-thiosemicarbazide
Hazard: Irritant
Formula:
C₈H₁₀ClN₃S Melting Pt: 174…175 °C |
MDL Number:
MFCD00060590 CAS Number: 6610-36-2 |
Specification Test Results
Melting point | 174-175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...