1-(4-Bromo-2-trifluoromethylphenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: 1-(4-Bromo-2-(trifluoromethyl)phenyl)thiourea

056933-1G 056933-5G
101911-784EA 90.77 USD
101911-784 101911-786
1-(4-Bromo-2-trifluoromethylphenyl)-2-thiourea
1-(4-Bromo-2-trifluoromethylphenyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₆BrF₃N₂S
Melting Pt: 178…180 °C
MDL Number: MFCD00041177
CAS Number: 208186-71-4

Order Now

Specification Test Results

Melting point 178-180°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR