1-(4-Bromo-2-methylphenyl)thiourea

Supplier: Matrix Scientific

Synonyms: N-(4-Bromo-2-methylphenyl)thiourea

056926-2G 056926-10G
101911-756EA 87.69 USD
101911-756 101911-758
1-(4-Bromo-2-methylphenyl)thiourea
1-(4-Bromo-2-methylphenyl)thiourea

Hazard: Irritant

Formula: C₈H₉BrN₂S
Melting Pt: 194…196 °C
MDL Number: MFCD00041178
CAS Number: 109317-23-9

Order Now

Specification Test Results

Melting point 194-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR