4-Bromo-3-methylbenzhydrazide

Supplier: Matrix Scientific

056920-2G 056920-10G
101911-734EA 153.85 USD
101911-734 101911-736
4-Bromo-3-methylbenzhydrazide
4-Bromo-3-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₉BrN₂O
MW: 229.08 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD00672945
CAS Number: 148672-43-9

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR