2-Bromo-4-methylbenzhydrazide

Supplier: Matrix Scientific

056918-2G 056918-10G
101911-726EA 126.15 USD
101911-726 101911-728
2-Bromo-4-methylbenzhydrazide
2-Bromo-4-methylbenzhydrazide

Hazard: Irritant

Formula: C₈H₉BrN₂O
MW: 229.08 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD03425674
CAS Number: 1022059-55-7

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR