3-Bromo-4-methoxybenzonitrile ≥98%

Supplier: Matrix Scientific

056916-5G 056916-25G 056916-100G
101911-716EA 123.08 USD
101911-716 101911-718 101911-720
3-Bromo-4-methoxybenzonitrile ≥98%
3-Bromo-4-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₆BrNO
Melting Pt: 122…124 °C
MDL Number: MFCD00143097
CAS Number: 117572-79-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 122-124°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR