1-Benzoyl-2-thiobiuret

Supplier: Matrix Scientific

056893-10G 056893-50G
101911-620EA 152.31 USD
101911-620 101911-622
1-Benzoyl-2-thiobiuret
1-Benzoyl-2-thiobiuret

Hazard: Irritant

Formula: C₉H₉N₃O₂S
Melting Pt: 173…175 °C
MDL Number: MFCD00025451
CAS Number: 41835-24-9

Order Now

Specification Test Results

Melting point 173-175°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR