3-Amino-4-methoxybenzanilide

Supplier: Matrix Scientific

056888-10G 056888-50G
101911-604EA 146.15 USD
101911-604 101911-606
3-Amino-4-methoxybenzanilide
3-Amino-4-methoxybenzanilide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
Melting Pt: 154…156 °C
MDL Number: MFCD00017166
CAS Number: 120-35-4

Order Now

Specification Test Results

Melting point 154-156°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR