2-Amino-N-methylbenzamide ≥98%

Supplier: Matrix Scientific

056638-5G 056638-25G
101911-106EA 21.54 USD
101911-106 101911-108
2-Amino-N-methylbenzamide ≥98%
2-Amino-N-methylbenzamide

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Melting Pt: 241-243°C
MDL Number: MFCD00060602
CAS Number: 4141-08-6

Order Now

Specification Test Results

Melting point 75-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR