5-Bromothiophene-2-carbohydrazide ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromothiophene-2-carboxylic acid hydrazide

056629-1G 056629-5G
101911-088EA 89.23 USD
101911-088 101911-090
5-Bromothiophene-2-carbohydrazide ≥98%
5-Bromothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₅H₅BrN₂OS
MW: 221.072 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD01310798
CAS Number: 98027-27-1

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR