1-(4-(Trifluoromethyl)phenyl)thiourea

Supplier: Matrix Scientific

Synonyms: N-[4-(Trifluoromethyl)phenyl]thiourea

056587-1G 056587-5G
101910-964EA 52.31 USD
101910-964 101910-966
1-(4-(Trifluoromethyl)phenyl)thiourea
1-(4-(Trifluoromethyl)phenyl)thiourea

Hazard: Irritant

Formula: C₈H₇F₃N₂S
Melting Pt: 144…145 °C
MDL Number: MFCD00041189
CAS Number: 1736-72-7

Order Now

Specification Test Results

Melting point 144-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR