4-[2-Chloro-5-(trifluoromethyl)phenyl]-3-thiosemicarbazide

Supplier: Matrix Scientific

056578-1G 056578-5G
101910-938EA 89.23 USD
101910-938 101910-940
4-[2-Chloro-5-(trifluoromethyl)phenyl]-3-thiosemicarbazide
4-[2-Chloro-5-(trifluoromethyl)phenyl]-3-thiosemicarbazide

Hazard: Irritant

Formula: C₈H₇ClF₃N₃S
Melting Pt: 156…158 °C
MDL Number: MFCD00041274
CAS Number: 206559-51-5

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR