Isothiocyanatophenyl sulfone

Supplier: Matrix Scientific

056573-1G 056573-5G
101910-920EA 69.23 USD
101910-920 101910-922
Isothiocyanatophenyl sulfone
Isothiocyanatophenyl sulfone

Hazard: Irritant-Harmful

Formula: C₁₄H₈N₂O₂S₃
Melting Pt: 187…189 °C
MDL Number: MFCD00041098
CAS Number: 4430-49-3

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR