4-Isothiocyanato-4'-nitrodiphenyl sulfide

Supplier: Matrix Scientific

056571-1G 056571-5G
101910-912EA 95.38 USD
101910-912 101910-914
4-Isothiocyanato-4'-nitrodiphenyl sulfide
4-Isothiocyanato-4'-nitrodiphenyl sulfide

Hazard: Irritant-Harmful

Formula: C₁₃H₈N₂O₂S₂
Melting Pt: 124…126 °C
MDL Number: MFCD00060547
CAS Number: 19822-35-6

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR