tert-Butyl-4-[2-(hydrazinecarbonyl)-4-nitrophenyl]piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

056512-500MG 056512-1G
101910-800EA 375.38 USD
101910-800 101910-802
tert-Butyl-4-[2-(hydrazinecarbonyl)-4-nitrophenyl]piperazine-1-carboxylate ≥95%
tert-Butyl-4-[2-(hydrazinecarbonyl)-4-nitrophenyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₃N₅O₅
MW: 365.39 g/mol
Melting Pt: 202…204 °C
MDL Number: MFCD16890132
CAS Number: 1242267-89-5

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR