tert-Butyl-4-(6-chloro-3-nitropyridin-2-yl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

056511-500MG 056511-1G
101910-796EA 261.54 USD
101910-796 101910-798
tert-Butyl-4-(6-chloro-3-nitropyridin-2-yl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(6-chloro-3-nitropyridin-2-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉ClN₄O₄
MW: 342.78 g/mol
Melting Pt: 94…95 °C
MDL Number: MFCD16890138
CAS Number: 474330-06-8

Order Now

Specification Test Results

Melting point 94-95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR