Pyrimidine-2-carboximidamide ≥95%

Supplier: Matrix Scientific

056507-500MG 056507-1G 056507-5G
101910-778EA 353.85 USD
101910-778 101910-780 101910-782
Pyrimidine-2-carboximidamide ≥95%
Pyrimidine-2-carboximidamide

Hazard: Irritant

Formula: C₅H₆N₄
MW: 122.13 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD09755980
CAS Number: 45695-56-5

Order Now

Specification Test Results

Melting point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR