1-Methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylic acid

056503-500MG 056503-1G
101910-764EA 769.23 USD
101910-764 101910-766
1-Methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid ≥95%
1-Methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₃
MW: 192.18 g/mol
Melting Pt: >350 °C
MDL Number: MFCD11131632
CAS Number: 19950-97-1

Order Now

Specification Test Results

Melting point >350°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR