Methyl-6-bromopyrazolo[1,5-a]pyrimidine-2-carboxylate ≥95%

Supplier: Matrix Scientific

056498-500MG 056498-1G
101910-746EA 512.31 USD
101910-746 101910-748
Methyl-6-bromopyrazolo[1,5-a]pyrimidine-2-carboxylate ≥95%
Methyl-6-bromopyrazolo[1,5-a]pyrimidine-2-carboxylate

Hazard: Irritant

Formula: C₈H₆BrN₃O₂
Melting Pt: 236…238 °C (dec.)
MDL Number: MFCD16890139
CAS Number: 1005209-40-4

Order Now

Specification Test Results

Melting point 236-238°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR