Methyl-4-aminothieno[2,3-d]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

056497-500MG 056497-1G
101910-742EA 556.92 USD
101910-742 101910-744
Methyl-4-aminothieno[2,3-d]pyrimidine-6-carboxylate ≥95%
Methyl-4-aminothieno[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₈H₇N₃O₂S
MW: 209.23 g/mol
Melting Pt: 266…268 °C
MDL Number: MFCD09842619
CAS Number: 155087-15-3

Order Now

Specification Test Results

Melting point 266-268°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR