Methyl-2-amino-5-bromoisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-bromo-4-pyridinecarboxylate

056494-500MG 056494-1G 056494-5G
101910-728EA 278.46 USD
101910-728 101910-730 101910-732
Methyl-2-amino-5-bromoisonicotinate ≥95%
Methyl 2-amino-5-bromoisonicotinate

Hazard: Irritant

Formula: C₇H₇BrN₂O₂
MW: 231.0467 g/mol
Melting Pt: 136-137°C
MDL Number: MFCD09027294
CAS Number: 882499-87-8

Order Now

Specification Test Results

Melting point 136-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR