1-(4-Iodophenyl)piperidin-2-one ≥95%

Supplier: Matrix Scientific

056492-500MG 056492-1G
101910-722EA 61.54 USD
101910-722 101910-724
1-(4-Iodophenyl)piperidin-2-one ≥95%
1-(4-Iodophenyl)piperidin-2-one

Hazard: Irritant

Formula: C₁₁H₁₂INO
Melting Pt: 118…120 °C
MDL Number: MFCD04037128
CAS Number: 385425-15-0

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR