Imidazo[1,2-a]pyridine-7-carboxylic acid ≥95%

Supplier: Matrix Scientific

056491-500MG 056491-1G
101910-718EA 107.69 USD
101910-718 101910-720
Imidazo[1,2-a]pyridine-7-carboxylic acid ≥95%
Imidazo[1,2-a]pyridine-7-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
Melting Pt: 295-296°C (dec)
MDL Number: MFCD09834781
CAS Number: 648423-85-2

Order Now

Specification Test Results

Melting point 295-296°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR