Ethyl-2-(diphenylamino)-4-methyl-1,3-thiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

056488-500MG 056488-1G 056488-5G
101910-702EA 375.38 USD
101910-702 101910-704 101910-706
Ethyl-2-(diphenylamino)-4-methyl-1,3-thiazole-5-carboxylate ≥95%
Ethyl-2-(diphenylamino)-4-methyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₉H₁₈N₂O₂S
Melting Pt: 155…157 °C
MDL Number: MFCD16890145
CAS Number: 1242267-87-3

Order Now

Specification Test Results

Melting point 155-157°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR