5,6-Dichloro-3-pyridinemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: (5,6-Dichloropyridin-3-yl)methanol, 2,3-Dichloro-5-hydroxymethylpyridine, 5,6-Dichloronicotinyl Alcohol

056477-1G 056477-5G
101910-652EA 106.15 USD
101910-652 101910-654
5,6-Dichloro-3-pyridinemethanol ≥95%
5,6-Dichloro-3-pyridinemethanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD00671515
CAS Number: 54127-30-9

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR