8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decane hydrochloride ≥95%

Supplier: Matrix Scientific

056473-500MG 056473-1G
101910-636EA 375.38 USD
101910-636 101910-638
8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decane hydrochloride ≥95%
8-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1-thia-4,8-diazaspiro[4.5]decane hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₆Cl₂F₃N₃S
Melting Pt: 175…176 °C
MDL Number: MFCD16890142
CAS Number: 1242267-83-9

Order Now

Specification Test Results

Melting point 175-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR