N-(1-Benzylpiperidin-4-yl)-3-chloro-5-(trifluoromethyl)pyridin-2-amine ≥95%

Supplier: Matrix Scientific

056468-500MG 056468-1G
101910-616EA 181.54 USD
101910-616 101910-618
N-(1-Benzylpiperidin-4-yl)-3-chloro-5-(trifluoromethyl)pyridin-2-amine ≥95%
N-(1-Benzylpiperidin-4-yl)-3-chloro-5-(trifluoromethyl)pyridin-2-amine

Hazard: Irritant

Formula: C₁₈H₁₉ClF₃N₃
Melting Pt: 63…65 °C
MDL Number: MFCD16890147
CAS Number: 1242267-94-2

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR